ENA321762427 Enamine

2-chloro-N-[(5-chloro-2-methoxyphenyl)methyl]acetamide

  • MDL number MFCD16486558
  • Cas Number : 1225915-88-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-48086

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-48086
Availability details for
Quantity
  • Availability
  • Add to Cart