ENA321762429 Enamine

1-(5-bromothiophen-2-yl)-2-phenylethan-1-one

  • MDL number MFCD02187511
  • Cas Number : 56824-81-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73796

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73796
Availability details for
Quantity
  • Availability
  • Add to Cart