ENA321762431 Enamine

N-cyclopentylpiperidin-4-amine

  • MDL number MFCD06740676
  • Cas Number : 886506-60-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73798

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73798
Availability details for
Quantity
  • Availability
  • Add to Cart