ENA321762434 Enamine

ethyl 6-(chlorosulfonyl)pyridine-3-carboxylate

  • MDL number MFCD16093665
  • Cas Number : 1249304-48-0
  • Molecular Weight : 249.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -70
Physical Form liquid
Shipping Temperature Dry Ice
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-73801

Safety Information

signalword Danger
pcodes P260,P261,P264,P270,P271,P280,P301+P312,P301+P330+P331,P302+P352,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P330,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-73801
pictograms GHS05,GHS07
hstatements H302,H312,H314,H332
Availability details for
Quantity
  • Availability
  • Add to Cart