ENA321762435 Enamine

N-[1-(1-benzofuran-2-yl)ethyl]-2-chloropropanamide

  • MDL number MFCD12496833
  • Cas Number : 1154010-39-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-28052

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28052
Availability details for
Quantity
  • Availability
  • Add to Cart