ENA321762436 Enamine

2-(2-fluorophenyl)-4,5,6,7-tetrahydro-1,3-benzothiazol-7-one

  • MDL number MFCD17437827
  • Cas Number : 1311315-49-7
  • Molecular Weight : 247.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-73803

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73803
Availability details for
Quantity
  • Availability
  • Add to Cart