ENA321762439 Enamine

methyl 3-(chlorosulfonyl)-1-benzothiophene-2-carboxylate

  • MDL number MFCD18483216
  • Cas Number : 74370-96-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98...100
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73806

Safety Information

msds https://www.enaminestore.com/catalog/EN300-73806
Availability details for
Quantity
  • Availability
  • Add to Cart