ENA321762442 Enamine

2-chloro-N-(3-cyclohexylpropyl)acetamide

  • MDL number MFCD18785438
  • Cas Number : 501008-00-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-48201

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-48201
Availability details for
Quantity
  • Availability
  • Add to Cart