ENA321762443 Enamine

2-(4-methoxyphenyl)-4,5,6,7-tetrahydro-1,3-benzothiazol-7-amine

  • MDL number MFCD16990730
  • Cas Number : 1251924-95-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73810

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73810
Availability details for
Quantity
  • Availability
  • Add to Cart