ENA321762448 Enamine

2-chloro-N-[1-(4-chlorophenyl)ethyl]propanamide

  • MDL number MFCD11643909
  • Cas Number : 92546-75-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-28057

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28057
Availability details for
Quantity
  • Availability
  • Add to Cart