ENA321762452 Enamine

1-(3-methylcyclohexyl)piperidin-4-one

  • MDL number MFCD12166449
  • Cas Number : 1096306-10-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73819

Safety Information

msds https://www.enaminestore.com/catalog/EN300-73819
Availability details for
Quantity
  • Availability
  • Add to Cart