ENA321762455 Enamine

1-(piperidin-4-yl)-4-propylpiperazine

  • MDL number MFCD09940719
  • Cas Number : 874099-63-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73825

Safety Information

msds https://www.enaminestore.com/catalog/EN300-73825
Availability details for
Quantity
  • Availability
  • Add to Cart