ENA321762457 Enamine

1-(3-fluorophenoxy)propan-2-ol

  • MDL number MFCD16102271
  • Cas Number : 2366-93-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-73827

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73827
Availability details for
Quantity
  • Availability
  • Add to Cart