ENA321762462 Enamine

2-chloro-N-(2-methanesulfonylphenyl)propanamide

  • MDL number MFCD11643910
  • Cas Number : 1098361-08-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28096

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28096
Availability details for
Quantity
  • Availability
  • Add to Cart