ENA321762465 Enamine

3-(2,5-dichlorophenyl)-1H-pyrazole

  • MDL number MFCD02091525
  • Cas Number : 59843-71-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 143...145
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73842

Safety Information

msds https://www.enaminestore.com/catalog/EN300-73842
Availability details for
Quantity
  • Availability
  • Add to Cart