ENA321762468 Enamine

2-chloro-N-ethyl-N-(2-methylprop-2-en-1-yl)propanamide

  • MDL number MFCD09942842
  • Cas Number : 1016790-17-2
  • Molecular Weight : 189.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-28124

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28124
Availability details for
Quantity
  • Availability
  • Add to Cart