ENA321762469 Enamine

2-chloro-N-methyl-N-[(2-methylphenyl)methyl]propanamide

  • MDL number MFCD12194364
  • Cas Number : 1156116-10-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-28149

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28149
Availability details for
Quantity
  • Availability
  • Add to Cart