ENA321762471 Enamine

1-(1-benzofuran-2-yl)-2-methylpropan-1-one

  • MDL number MFCD11203483
  • Cas Number : 91420-41-6
  • Molecular Weight : 188.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-73849

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73849
Availability details for
Quantity
  • Availability
  • Add to Cart