ENA321762474 Enamine

5-methyl-3-phenyl-1,3-diazinane-2,4-dione

  • MDL number MFCD16839962
  • Cas Number : 85606-74-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 174...176
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73853

Safety Information

msds https://www.enaminestore.com/catalog/EN300-73853
Availability details for
Quantity
  • Availability
  • Add to Cart