ENA321762476 Enamine

2-chloro-N-[1-oxo-1-(piperidin-1-yl)propan-2-yl]acetamide

  • MDL number MFCD01497682
  • Cas Number : 1218022-97-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-48392

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-48392
Availability details for
Quantity
  • Availability
  • Add to Cart