ENA321762477 Enamine

1-benzofuran-2-carbothioamide

  • MDL number MFCD11179436
  • Cas Number : 190430-18-3
  • Molecular Weight : 177.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-73858

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73858
Availability details for
Quantity
  • Availability
  • Add to Cart