ENA321762481 Enamine

1-[4-(trifluoromethyl)phenyl]-2,3-dihydro-1H-imidazole-2-thione

  • MDL number MFCD00060485
  • Cas Number : 17452-18-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73868

Safety Information

msds https://www.enaminestore.com/catalog/EN300-73868
Availability details for
Quantity
  • Availability
  • Add to Cart