ENA321762483 Enamine

2-(1-benzofuran-2-yl)-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD11102965
  • Cas Number : 1152604-97-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73870

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73870
Availability details for
Quantity
  • Availability
  • Add to Cart