ENA321762492 Enamine

N-(4-hydroxyphenyl)cyclohexanecarboxamide

  • MDL number MFCD02674231
  • Cas Number : 548763-46-8
  • Molecular Weight : 219.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-73881

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73881
Availability details for
Quantity
  • Availability
  • Add to Cart