ENA321762496 Enamine

2-(3-ethylpiperidin-1-yl)-2-methylpropan-1-amine

  • MDL number MFCD12143219
  • Cas Number : 1156659-65-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-73887

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73887
Availability details for
Quantity
  • Availability
  • Add to Cart