ENA321762499 Enamine

2-[2-(propan-2-yloxy)ethyl]piperidine hydrochloride

  • MDL number MFCD13560728
  • Cas Number : 1219972-32-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73892

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73892
Availability details for
Quantity
  • Availability
  • Add to Cart