ENA321762503 Enamine

2-({1-[4-(trifluoromethyl)phenyl]-1H-imidazol-2-yl}sulfanyl)acetic acid

  • MDL number MFCD13465105
  • Cas Number : 1038335-12-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-73912

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73912
Availability details for
Quantity
  • Availability
  • Add to Cart