ENA321762506 Enamine

5,6-dimethyl-7-(pyridin-2-ylmethyl)-7H-pyrrolo[2,3-d]pyrimidin-4-amine

  • MDL number MFCD12653380
  • Cas Number : 1096957-99-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73915

Safety Information

msds https://www.enaminestore.com/catalog/EN300-73915
Availability details for
Quantity
  • Availability
  • Add to Cart