ENA321762521 Enamine

N-(3-amino-4-methylphenyl)-2,2-dimethylpropanamide

  • MDL number MFCD09931546
  • Cas Number : 946709-89-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73937

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73937
Availability details for
Quantity
  • Availability
  • Add to Cart