ENA321762525 Enamine

benzyl[(oxan-4-yl)methyl]amine

  • MDL number MFCD07371441
  • Cas Number : 887978-20-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-73942

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73942
Availability details for
Quantity
  • Availability
  • Add to Cart