ENA321762530 Enamine

N-(3-aminopropyl)-3-fluorobenzamide hydrochloride

  • MDL number MFCD18483237
  • Cas Number : 1280980-98-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73953

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73953
Availability details for
Quantity
  • Availability
  • Add to Cart