ENA321762531 Enamine

2-(pyridin-4-yloxy)aniline

  • MDL number MFCD00956807
  • Molecular Weight : 186.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-73954

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73954
Availability details for
Quantity
  • Availability
  • Add to Cart