ENA321762532 Enamine

2-amino-N-[2-(propan-2-yl)-1,3-benzoxazol-5-yl]butanamide

  • MDL number MFCD12703860
  • Cas Number : 1218287-66-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73957

Safety Information

msds https://www.enaminestore.com/catalog/EN300-73957
Availability details for
Quantity
  • Availability
  • Add to Cart