ENA321762562 Enamine

[2-(pyridin-3-yl)ethyl]urea

  • MDL number MFCD12798242
  • Cas Number : 1179193-35-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74002

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74002
Availability details for
Quantity
  • Availability
  • Add to Cart