ENA321762576 Enamine

2-ethyl-1,3-thiazole-5-carbaldehyde

  • MDL number MFCD14588408
  • Cas Number : 933683-87-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51448

Safety Information

msds https://www.enaminestore.com/catalog/EN300-51448
Availability details for
Quantity
  • Availability
  • Add to Cart