ENA321762579 Enamine

3-methyl-[1,2]oxazolo[5,4-b]pyridine-5-carbohydrazide

  • MDL number MFCD11644000
  • Cas Number : 1097794-89-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-74104

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74104
Availability details for
Quantity
  • Availability
  • Add to Cart