ENA321762587 Enamine

methyl 3-{[(3,4-difluorophenyl)methyl]amino}propanoate

  • MDL number MFCD11102609
  • Cas Number : 1152548-66-7
  • Molecular Weight : 229.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-74118

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74118
Availability details for
Quantity
  • Availability
  • Add to Cart