ENA321762588 Enamine

[4-(2-methoxyphenoxy)phenyl]methanamine

  • MDL number MFCD07365254
  • Cas Number : 893752-93-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74119

Safety Information

msds https://www.enaminestore.com/catalog/EN300-74119
Availability details for
Quantity
  • Availability
  • Add to Cart