ENA321762594 Enamine

7-methyl-1,2,3,4-tetrahydroquinoline

  • MDL number MFCD07658380
  • Cas Number : 58960-03-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74126

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74126
Availability details for
Quantity
  • Availability
  • Add to Cart