ENA321762595 Enamine

3-(2-fluorophenyl)-1-(1,2,3,4-tetrahydroquinolin-5-yl)urea

  • MDL number MFCD09809426
  • Cas Number : 1016498-13-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-74128

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74128
Availability details for
Quantity
  • Availability
  • Add to Cart