ENA321762597 Enamine

2-[4-(2-amino-4-chlorophenyl)piperazin-1-yl]ethan-1-ol

  • MDL number MFCD09163758
  • Cas Number : 946812-10-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-74130

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74130
Availability details for
Quantity
  • Availability
  • Add to Cart