ENA321762601 Enamine

5-(2-fluorophenyl)-1-methyl-1H-pyrrole-2-carboxylic acid

  • MDL number MFCD10006418
  • Cas Number : 1017414-92-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-74135
Availability details for
Quantity
  • Availability
  • Add to Cart