ENA321762603 Enamine

2-(4-aminophenoxy)ethan-1-ol

  • MDL number MFCD00025329
  • Cas Number : 6421-88-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-74137

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74137
Availability details for
Quantity
  • Availability
  • Add to Cart