ENA321762608 Enamine

methyl 1-(5-nitrothiophene-2-carbonyl)pyrrolidine-2-carboxylate

  • MDL number MFCD12644126

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-74143

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74143
Availability details for
Quantity
  • Availability
  • Add to Cart