ENA321762609 Enamine

2-(pyridin-4-yl)aniline

  • MDL number MFCD06738704
  • Cas Number : 106047-18-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 81-86
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74144

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74144
Availability details for
Quantity
  • Availability
  • Add to Cart