ENA321762611 Enamine

6-ethylpyrimidin-4-ol

  • MDL number MFCD05864423
  • Cas Number : 124703-78-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135...137
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53291

Safety Information

msds https://www.enaminestore.com/catalog/EN300-53291
Availability details for
Quantity
  • Availability
  • Add to Cart