ENA321762614 Enamine

methyl 1-(5-nitrofuran-2-carbonyl)pyrrolidine-2-carboxylate

  • MDL number MFCD12488690
  • Molecular Weight : 268.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-74149

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74149
Availability details for
Quantity
  • Availability
  • Add to Cart