ENA321762622 Enamine

5-[(4-bromophenyl)methyl]-3-(chloromethyl)-1,2,4-oxadiazole

  • MDL number MFCD10694678
  • Cas Number : 1152504-39-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74164

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74164
Availability details for
Quantity
  • Availability
  • Add to Cart