ENA321762626 Enamine

1-amino-3-methylcyclohexane-1-carboxylic acid

  • MDL number MFCD01745645
  • Cas Number : 55550-84-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-53603

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53603
Availability details for
Quantity
  • Availability
  • Add to Cart