ENA321762630 Enamine

N-(4-{[1-(2-methyl-1,3-thiazol-4-yl)ethyl]amino}phenyl)cyclopropanecarboxamide

  • MDL number MFCD13763357
  • Cas Number : 1156718-25-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74173

Safety Information

msds https://www.enaminestore.com/catalog/EN300-74173
Availability details for
Quantity
  • Availability
  • Add to Cart